5-[(2,4-Difluorophenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077405-500MG 077405-1G 077405-5G
102666-066EA 387.69 USD
102666-066 102666-068 102666-070
5-[(2,4-Difluorophenoxy)methyl]-2-furoic acid
5-[(2,4-Difluorophenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₂H₈F₂O₄
MDL Number: MFCD03421872
CAS Number: 832740-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR