5-[(4-Ethoxyphenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077404-500MG 077404-1G 077404-5G
102666-060EA 387.69 USD
102666-060 102666-062 102666-064
5-[(4-Ethoxyphenoxy)methyl]-2-furoic acid
5-[(4-Ethoxyphenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₄H₁₄O₅
MDL Number: MFCD02605780
CAS Number: 438474-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR