5-[(2,3-Dichlorophenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077403-500MG 077403-1G 077403-5G
102666-054EA 387.69 USD
102666-054 102666-056 102666-058
5-[(2,3-Dichlorophenoxy)methyl]-2-furoic acid
5-[(2,3-Dichlorophenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₂H₈Cl₂O₄
MDL Number: MFCD03421873
CAS Number: 438218-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR