5-[(4-Chloro-2-isopropyl-5-methylphenoxy)methyl]-2-furoic acid
Supplier: Matrix Scientific
077401-500MG
077401-1G
077401-5G
102666-042EA
387.69
USD
102666-042
102666-044
102666-046
5-[(4-Chloro-2-isopropyl-5-methylphenoxy)methyl]-2-furoic acid
5-[(4-Chloro-2-isopropyl-5-methylphenoxy)methyl]-2-furoic acid
Hazard: Irritant
Formula:
C₁₆H₁₇ClO₄ MW: 308.76 g/mol |
MDL Number:
MFCD03074360 CAS Number: 438219-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...