5-[(3-Formylphenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077400-500MG 077400-1G 077400-5G
102666-036EA 387.69 USD
102666-036 102666-038 102666-040
5-[(3-Formylphenoxy)methyl]-2-furoic acid
5-[(3-Formylphenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₃H₁₀O₅
MDL Number: MFCD06653194
CAS Number: 881041-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR