5-[(4-Ethylphenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077399-500MG 077399-1G 077399-5G
102666-030EA 387.69 USD
102666-030 102666-032 102666-034
5-[(4-Ethylphenoxy)methyl]-2-furoic acid
5-[(4-Ethylphenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₄H₁₄O₄
MDL Number: MFCD02253884
CAS Number: 364595-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR