5-[(2,4,6-Trichlorophenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077398-500MG 077398-1G 077398-5G
102666-024EA 258.46 USD
102666-024 102666-026 102666-028
5-[(2,4,6-Trichlorophenoxy)methyl]-2-furoic acid
5-[(2,4,6-Trichlorophenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₂H₇Cl₃O₄
MDL Number: MFCD03419435
CAS Number: 438220-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR