5-[(2-Chloro-6-methylphenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077397-500MG 077397-1G 077397-5G
102666-018EA 387.69 USD
102666-018 102666-020 102666-022
5-[(2-Chloro-6-methylphenoxy)methyl]-2-furoic acid
5-[(2-Chloro-6-methylphenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₃H₁₁ClO₄
MDL Number: MFCD03421883
CAS Number: 832737-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR