5-[(2,4-Dimethylphenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077396-500MG 077396-1G 077396-5G
102666-012EA 387.69 USD
102666-012 102666-014 102666-016
5-[(2,4-Dimethylphenoxy)methyl]-2-furoic acid
5-[(2,4-Dimethylphenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₄H₁₄O₄
MDL Number: MFCD03421880
CAS Number: 869950-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR