3-[(2,3-Dichlorophenoxy)methyl]-4-methoxybenzoic acid

Supplier: Matrix Scientific

077394-500MG 077394-1G 077394-5G
102666-000EA 258.46 USD
102666-000 102666-002 102666-004
3-[(2,3-Dichlorophenoxy)methyl]-4-methoxybenzoic acid
3-[(2,3-Dichlorophenoxy)methyl]-4-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₂Cl₂O₄
MDL Number: MFCD03419430
CAS Number: 438220-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR