3,5-Dimethyl-4-[(4-nitrophenoxy)methyl]isoxazole
Supplier: Matrix Scientific
077387-500MG
077387-1G
077387-5G
102665-960EA
387.69
USD
102665-960
102665-962
102665-964
3,5-Dimethyl-4-[(4-nitrophenoxy)methyl]isoxazole
3,5-Dimethyl-4-[(4-nitrophenoxy)methyl]isoxazole
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₄ |
MDL Number:
MFCD10485467 CAS Number: 951923-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...