[(4-Amino-4H-1,2,4-triazol-3-yl)thio]acetonitrile

Supplier: Matrix Scientific

077377-500MG 077377-1G 077377-5G
102665-902EA 516.92 USD
102665-902 102665-904 102665-906
[(4-Amino-4H-1,2,4-triazol-3-yl)thio]acetonitrile
[(4-Amino-4H-1,2,4-triazol-3-yl)thio]acetonitrile

Hazard: Irritant

Formula: C₄H₅N₅S
MDL Number: MFCD06803473
CAS Number: 74599-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR