3-Propyl[1,2,4]triazolo[3,4-b][1,3,4]thiadiazol-6-amine
Supplier: Matrix Scientific
077374-500MG
077374-1G
077374-5G
102665-884EA
387.69
USD
102665-884
102665-886
102665-888
3-Propyl[1,2,4]triazolo[3,4-b][1,3,4]thiadiazol-6-amine
3-Propyl[1,2,4]triazolo[3,4-b][1,3,4]thiadiazol-6-amine
Hazard: Irritant
Formula:
C₆H₉N₅S |
MDL Number:
MFCD06803569 CAS Number: 217788-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...