3-(Difluoromethyl)-5-(methylthio)-4H-1,2,4-triazol-4-amine

Supplier: Matrix Scientific

077373-500MG 077373-1G 077373-5G
102665-878EA 387.69 USD
102665-878 102665-880 102665-882
3-(Difluoromethyl)-5-(methylthio)-4H-1,2,4-triazol-4-amine
3-(Difluoromethyl)-5-(methylthio)-4H-1,2,4-triazol-4-amine

Hazard: Irritant

Formula: C₄H₆F₂N₄S
MDL Number: MFCD04969127
CAS Number: 869942-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR