Methyl-2-{[4-(chlorosulfonyl)butyl]amino}benzoate

Supplier: Matrix Scientific

077370-500MG 077370-1G 077370-5G
102665-860EA 193.85 USD
102665-860 102665-862 102665-864
Methyl-2-{[4-(chlorosulfonyl)butyl]amino}benzoate
Methyl-2-{[4-(chlorosulfonyl)butyl]amino}benzoate

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₄S
MW: 305.78 g/mol
MDL Number: MFCD20441348
CAS Number: 1338494-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR