4-(Pyridin-2-ylmethyl)piperazin-1-amine dihydrochloride
Supplier: Matrix Scientific
077369-500MG
077369-1G
077369-5G
102665-854EA
516.92
USD
102665-854
102665-856
102665-858
4-(Pyridin-2-ylmethyl)piperazin-1-amine dihydrochloride
4-(Pyridin-2-ylmethyl)piperazin-1-amine dihydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₈Cl₂N₄ |
MDL Number:
MFCD20441441 CAS Number: 1401425-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...