5-{[3-(Trifluoromethyl)phenoxy]-methyl}-2-furoic acid
Supplier: Matrix Scientific
077360-500MG
077360-1G
077360-5G
102665-800EA
258.46
USD
102665-800
102665-802
102665-804
5-{[3-(Trifluoromethyl)phenoxy]-methyl}-2-furoic acid
5-{[3-(Trifluoromethyl)phenoxy]-methyl}-2-furoic acid
Hazard: Irritant
Formula:
C₁₃H₉F₃O₄ |
MDL Number:
MFCD03421874 CAS Number: 74556-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...