5-[(4-Acetylphenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077358-500MG 077358-1G 077358-5G
102665-788EA 258.46 USD
102665-788 102665-790 102665-792
5-[(4-Acetylphenoxy)methyl]-2-furoic acid
5-[(4-Acetylphenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₅
MDL Number: MFCD03421882
CAS Number: 832740-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR