Ethyl-4-(5-fluoro-2-methoxyphenyl)-2,4-dioxobutanoate

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(5-fluoro-2-methoxyphenyl)-2,4-dioxobutanoate

077352-500MG 077352-1G 077352-5G
102665-752EA 258.46 USD
102665-752 102665-754 102665-756
Ethyl-4-(5-fluoro-2-methoxyphenyl)-2,4-dioxobutanoate
Ethyl-4-(5-fluoro-2-methoxyphenyl)-2,4-dioxobutanoate

Hazard: Irritant

Formula: C₁₃H₁₃FO₅
MW: 268.24 g/mol
MDL Number: MFCD16338212
CAS Number: 1225574-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR