4-Methoxy-3-[(quinolin-7-yloxy)methyl]benzaldehyde
Supplier: Matrix Scientific
077351-500MG
077351-1G
102665-748EA
258.46
USD
102665-748
102665-750
4-Methoxy-3-[(quinolin-7-yloxy)methyl]benzaldehyde
4-Methoxy-3-[(quinolin-7-yloxy)methyl]benzaldehyde
Hazard: Irritant
Formula:
C₁₈H₁₅NO₃ |
MDL Number:
MFCD20441346 CAS Number: 1338495-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...