4-(Chloromethyl)-1-isopropyl-1H-pyrazole hydrochloride
Supplier: Matrix Scientific
077345-500MG
077345-1G
077345-5G
102665-712EA
516.92
USD
102665-712
102665-714
102665-716
4-(Chloromethyl)-1-isopropyl-1H-pyrazole hydrochloride
4-(Chloromethyl)-1-isopropyl-1H-pyrazole hydrochloride
Hazard: Irritant
Formula:
C₇H₁₂Cl₂N₂ |
MDL Number:
MFCD20441440 CAS Number: 1401425-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...