1-Ethyl-1H-pyrazole-4-carbonitrile

Supplier: Matrix Scientific

077340-500MG 077340-1G 077340-5G
102665-684EA 516.92 USD
102665-684 102665-686 102665-688
1-Ethyl-1H-pyrazole-4-carbonitrile
1-Ethyl-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₆H₇N₃
MDL Number: MFCD04969705
CAS Number: 1006434-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR