3,5-Dibromo-1-(tetrahydro-2H-pyran-2-yl)-1H-1,2,4-triazole

Supplier: Matrix Scientific

077335-500MG 077335-1G
102665-660EA 387.69 USD
102665-660 102665-662
3,5-Dibromo-1-(tetrahydro-2H-pyran-2-yl)-1H-1,2,4-triazole
3,5-Dibromo-1-(tetrahydro-2H-pyran-2-yl)-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₇H₉Br₂N₃O
MDL Number: MFCD03931111
CAS Number: 106724-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR