Ethyl (3-hydroxy-1H-pyrazol-4-yl)acetate

Supplier: Matrix Scientific

077333-500MG 077333-1G
102665-654EA 455.38 USD
102665-654 102665-656
Ethyl (3-hydroxy-1H-pyrazol-4-yl)acetate
Ethyl (3-hydroxy-1H-pyrazol-4-yl)acetate

Hazard: Irritant

Formula: C₇H₁₀N₂O₃
MDL Number: MFCD09034162
CAS Number: 890095-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR