5-sec-Butyl-4-hydroxyisophthalic acid

Supplier: Matrix Scientific

077331-500MG 077331-1G
102665-646EA 387.69 USD
102665-646 102665-648
5-sec-Butyl-4-hydroxyisophthalic acid
5-sec-Butyl-4-hydroxyisophthalic acid

Hazard: Irritant

Formula: C₁₂H₁₄O₅
MDL Number: MFCD02925616
CAS Number: 13253-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR