5,5-Bis(hydroxymethyl)-1,3-thiazolidine-2,4-dione

Supplier: Matrix Scientific

077326-500MG 077326-1G 077326-5G
102665-620EA 258.46 USD
102665-620 102665-622 102665-624
5,5-Bis(hydroxymethyl)-1,3-thiazolidine-2,4-dione
5,5-Bis(hydroxymethyl)-1,3-thiazolidine-2,4-dione

Hazard: Irritant

Formula: C₅H₇NO₄S
MDL Number: MFCD01243276
CAS Number: 353467-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR