3-Phenylquinuclidin-3-ol hydrochloride

Supplier: Matrix Scientific

077322-500MG 077322-1G 077322-5G
102665-598EA 387.69 USD
102665-598 102665-600 102665-602
3-Phenylquinuclidin-3-ol hydrochloride
3-Phenylquinuclidin-3-ol hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO
MDL Number: MFCD03869032
CAS Number: 107150-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR