1-(1,3-Benzodioxol-5-ylmethyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

077317-500MG 077317-1G
102665-570EA 541.54 USD
102665-570 102665-572
1-(1,3-Benzodioxol-5-ylmethyl)-5-oxopyrrolidine-3-carboxylic acid
1-(1,3-Benzodioxol-5-ylmethyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₃NO₅
MDL Number: MFCD01330896
CAS Number: 261363-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR