1-(2-Naphthyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

077316-500MG 077316-1G
102665-566EA 258.46 USD
102665-566 102665-568
1-(2-Naphthyl)-5-oxopyrrolidine-3-carboxylic acid
1-(2-Naphthyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₃NO₃
MDL Number: MFCD08133402
CAS Number: 566154-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR