Methyl-3-(2-oxo-1,3-benzoxazol-3(2H)-yl)propanoate
Supplier: Matrix Scientific
077312-500MG
077312-1G
102665-546EA
387.69
USD
102665-546
102665-548
Methyl-3-(2-oxo-1,3-benzoxazol-3(2H)-yl)propanoate
Methyl-3-(2-oxo-1,3-benzoxazol-3(2H)-yl)propanoate
Hazard: Irritant
Formula:
C₁₁H₁₁NO₄ |
MDL Number:
MFCD11175284 CAS Number: 28884-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...