2-(3-Phenylpyrrolidin-1-yl)acetohydrazide
Supplier: Matrix Scientific
077311-500MG
077311-1G
102665-542EA
516.92
USD
102665-542
102665-544
2-(3-Phenylpyrrolidin-1-yl)acetohydrazide
2-(3-Phenylpyrrolidin-1-yl)acetohydrazide
Hazard: Irritant
Formula:
C₁₂H₁₇N₃O |
MDL Number:
MFCD18370861 CAS Number: 1313825-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...