(3-Phenylpyrrolidin-1-yl)acetic acid

Supplier: Matrix Scientific

077310-500MG 077310-1G
102665-538EA 516.92 USD
102665-538 102665-540
(3-Phenylpyrrolidin-1-yl)acetic acid
(3-Phenylpyrrolidin-1-yl)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD09971152
CAS Number: 1083299-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR