Methyl-3-(1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)propanoate
Supplier: Matrix Scientific
077309-500MG
077309-1G
102665-534EA
387.69
USD
102665-534
102665-536
Methyl-3-(1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)propanoate
Methyl-3-(1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)propanoate
Hazard: Irritant
Formula:
C₁₁H₁₄N₄O₄ |
MDL Number:
MFCD01417777 CAS Number: 331972-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...