8-Bromo-7-isopropyl-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione

Supplier: Matrix Scientific

077307-500MG 077307-1G
102665-526EA 387.69 USD
102665-526 102665-528
8-Bromo-7-isopropyl-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione
8-Bromo-7-isopropyl-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione

Hazard: Irritant

Formula: C₁₀H₁₃BrN₄O₂
MDL Number: MFCD00687948
CAS Number: 368840-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR