4-(4-Methylpiperidin-1-yl)butanoic acid
Supplier: Matrix Scientific
077305-500MG
077305-1G
077305-5G
102665-514EA
923.08
USD
102665-514
102665-516
102665-518
4-(4-Methylpiperidin-1-yl)butanoic acid
4-(4-Methylpiperidin-1-yl)butanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₉NO₂ |
MDL Number:
MFCD09710614 CAS Number: 775248-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...