4-[Benzyl(methyl)amino]butanoic acid

Supplier: Matrix Scientific

077304-500MG 077304-1G
102665-510EA 516.92 USD
102665-510 102665-512
4-[Benzyl(methyl)amino]butanoic acid
4-[Benzyl(methyl)amino]butanoic acid

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD09704258
CAS Number: 188132-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR