2-[Benzyl(methyl)amino]acetohydrazide
Supplier: Matrix Scientific
077302-500MG
077302-1G
102665-500EA
516.92
USD
102665-500
102665-502
2-[Benzyl(methyl)amino]acetohydrazide
2-[Benzyl(methyl)amino]acetohydrazide
Hazard: Irritant
Formula:
C₁₀H₁₅N₃O |
MDL Number:
MFCD15209120 CAS Number: 71480-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...