2-Azepan-1-ylacetohydrazide

Supplier: Matrix Scientific

077300-500MG 077300-1G 077300-5G
102665-488EA 516.92 USD
102665-488 102665-490 102665-492
2-Azepan-1-ylacetohydrazide
2-Azepan-1-ylacetohydrazide

Hazard: Irritant

Formula: C₈H₁₇N₃O
Melting Pt: 172…174 °C
MDL Number: MFCD08273568
CAS Number: 7408-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR