3-(Trifluoromethyl)[1,2,4]triazolo[3,4-b][1,3,4]thiadiazol-6-amine
Supplier: Matrix Scientific
077293-500MG
077293-1G
102665-452EA
516.92
USD
102665-452
102665-454
3-(Trifluoromethyl)[1,2,4]triazolo[3,4-b][1,3,4]thiadiazol-6-amine
3-(Trifluoromethyl)[1,2,4]triazolo[3,4-b][1,3,4]thiadiazol-6-amine
Hazard: Irritant
Formula:
C₄H₂F₃N₅S |
MDL Number:
MFCD06803573 CAS Number: 24848-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...