5-(1H-Benzimidazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077292-500MG 077292-1G 077292-5G
102665-446EA 387.69 USD
102665-446 102665-448 102665-450
5-(1H-Benzimidazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
5-(1H-Benzimidazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₀H₉N₅S
MDL Number: MFCD11868890
CAS Number: 1093854-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR