1-(Bromomethyl)propyl ethyl ether

Supplier: Matrix Scientific

077282-500MG 077282-1G 077282-5G
102665-388EA 369.23 USD
102665-388 102665-390 102665-392
1-(Bromomethyl)propyl ethyl ether
1-(Bromomethyl)propyl ethyl ether

Hazard: Irritant

Formula: C₆H₁₃BrO
Boiling Pt: 154…156 °C
Density: 1.2355
MDL Number: MFCD20441341
CAS Number: 34264-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR