N-[(1-Morpholin-4-ylcyclohexyl)methyl]benzamide
Supplier: Matrix Scientific
077280-500MG
077280-1G
102665-378EA
193.85
USD
102665-378
102665-380
N-[(1-Morpholin-4-ylcyclohexyl)methyl]benzamide
N-[(1-Morpholin-4-ylcyclohexyl)methyl]benzamide
Hazard: Irritant
Formula:
C₁₈H₂₆N₂O₂ |
MDL Number:
MFCD08545037 CAS Number: 939765-17-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...