2-Morpholin-4-yl-2-phenylpropanenitrile

Supplier: Matrix Scientific

077279-500MG 077279-1G 077279-5G
102665-372EA 387.69 USD
102665-372 102665-374 102665-376
2-Morpholin-4-yl-2-phenylpropanenitrile
2-Morpholin-4-yl-2-phenylpropanenitrile

Hazard: Irritant

Formula: C₁₃H₁₆N₂O
MDL Number: MFCD19350480
CAS Number: 477531-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR