3-(Trifluoromethyl)-1,2,4-oxadiazol-5-ol

Supplier: Matrix Scientific

077260-500MG 077260-1G 077260-5G
102665-276EA 646.15 USD
102665-276 102665-278 102665-280
3-(Trifluoromethyl)-1,2,4-oxadiazol-5-ol
3-(Trifluoromethyl)-1,2,4-oxadiazol-5-ol

Hazard: Irritant

Formula: C₃HF₃N₂O₂
MDL Number: MFCD20441336
CAS Number: 1338494-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR