2-(1H-Imidazol-2-ylmethyl)-1H-isoindole-1,3(2H)-dione
Supplier: Matrix Scientific
077253-500MG
077253-1G
102665-238EA
387.69
USD
102665-238
102665-240
2-(1H-Imidazol-2-ylmethyl)-1H-isoindole-1,3(2H)-dione
2-(1H-Imidazol-2-ylmethyl)-1H-isoindole-1,3(2H)-dione
Hazard: Irritant
Formula:
C₁₂H₉N₃O₂ |
MDL Number:
MFCD20180726 CAS Number: 760130-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...