3-(Bromomethyl)-1-ethylpyrrolidine hydrobromide

Supplier: Matrix Scientific

077242-500MG 077242-1G
102665-190EA 646.15 USD
102665-190 102665-192
3-(Bromomethyl)-1-ethylpyrrolidine hydrobromide
3-(Bromomethyl)-1-ethylpyrrolidine hydrobromide

Hazard: Irritant

Formula: C₇H₁₅Br₂N
MDL Number: MFCD20441432
CAS Number: 1390655-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR