2-Pyrrolidin-2-yl-1,3-benzoxazole

Supplier: Matrix Scientific

077231-500MG 077231-1G 077231-5G
102665-138EA 516.92 USD
102665-138 102665-140 102665-142
2-Pyrrolidin-2-yl-1,3-benzoxazole
2-Pyrrolidin-2-yl-1,3-benzoxazole

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD06660811
CAS Number: 885461-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR