[1-Methyl-2-(4-methylpiperazin-1-yl)ethyl]amine dihydrochloride

Supplier: Matrix Scientific

077225-500MG 077225-1G 077225-5G
102665-108EA 516.92 USD
102665-108 102665-110 102665-112
[1-Methyl-2-(4-methylpiperazin-1-yl)ethyl]amine dihydrochloride
[1-Methyl-2-(4-methylpiperazin-1-yl)ethyl]amine dihydrochloride

Hazard: Irritant

Formula: C₈H₂₁Cl₂N₃
MDL Number: MFCD20441464
CAS Number: 1401425-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR