[(2E)-1-Methyl-3-phenylprop-2-en-1-yl]amine hydrochloride

Supplier: Matrix Scientific

077222-500MG 077222-1G 077222-5G
102665-090EA 516.92 USD
102665-090 102665-092 102665-094
[(2E)-1-Methyl-3-phenylprop-2-en-1-yl]amine hydrochloride
[(2E)-1-Methyl-3-phenylprop-2-en-1-yl]amine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClN
MDL Number: MFCD01736902
CAS Number: 1356494-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR