7-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine hydrochloride
Supplier: Matrix Scientific
077221-500MG
077221-1G
077221-5G
102665-084EA
646.15
USD
102665-084
102665-086
102665-088
7-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine hydrochloride
7-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine hydrochloride
Hazard: Irritant
Formula:
C₆H₈ClN₅ |
MDL Number:
MFCD20441452 CAS Number: 1390654-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...